ContactUsLinkComponent
Gas Dosing Controller, Controls the inlet of gas into the SICCO Glove Box to reduce gas consumption. The set consists of a gas dosin g controller, a sensor cable, a tubing for connecting the flow meter with the measuring instrument and a power cable. 1 * 1 items
Catalog # BOHLV1775-02
Supplier:  Bohlender
undefined
Gas Dosing Controller, Controls the inlet of gas into the SICCO Glove Box to reduce gas consumption. The set consists of a gas dosin g controller, a sensor cable, a tubing for connecting the flow meter with the measuring instrument and a power cable. 1 * 1 items
Catalog # BOHLV1775-02
Supplier:  Bohlender
Frequently Bought Together