ContactUsLinkComponent
Magnetic Stirring Bars with Blade, PTFE-encapsulated magnetic core (Alnico 5), tapered blade with fixed cylindrical stirring bar, un iversal chemical resistance. 1 * 1 items
Catalog # BOHLC349-06
Supplier:  Bohlender
undefined
Magnetic Stirring Bars with Blade, PTFE-encapsulated magnetic core (Alnico 5), tapered blade with fixed cylindrical stirring bar, un iversal chemical resistance. 1 * 1 items
Catalog # BOHLC349-06
Supplier:  Bohlender
Frequently Bought Together