Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample Ejection Rods

Sample Ejection Rods

Sample ejection rod, Stainless steel 316L, 1100 mm
N° de catalogue SAMP8570A-1100
Fournisseur:   Sampling Systems
Sample Ejection Rods
N° de catalogue SAMP8570A-1100
Fournisseur:  Sampling Systems
Sample Ejection Rods
Sample Ejection Rods

Sample ejection rod, Stainless steel 316L, 1100 mm

Prix
250,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sample ejection rods are designed to facilitate the removal of samples.

  • A useful tool for removing samples
  • Choose a rod longer than the sampler being used
  • Available from stock
  • Fast delivery

The sample ejection rod is a convenient tool for removing any hard-to-eject samples collected within a sampler. Simply push the rod down the sampler shaft to eject the material.

Specifications
  • Description:
    Sample ejection rod
  • Length:
    1100 mm
  • Material:
    Stainless steel 316L
  • Seq:
    02