ContactUsLinkComponent
TemperatureProbes PT 1000, One measuring sensor PT 1000 in a PTFE-encapsulated stainless steel tube (1.4571). Temperatureprobe Ø 8m m, tip Ø 6mm, collar ring Ø 12mm. With white PFA-coated cable (length: 1,5m, 2-wire system). 1 * 1 items
Catalog # BOHLP1950-25
Supplier:  Bohlender
undefined
TemperatureProbes PT 1000, One measuring sensor PT 1000 in a PTFE-encapsulated stainless steel tube (1.4571). Temperatureprobe Ø 8m m, tip Ø 6mm, collar ring Ø 12mm. With white PFA-coated cable (length: 1,5m, 2-wire system). 1 * 1 items
Catalog # BOHLP1950-25
Supplier:  Bohlender
Frequently Bought Together