Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,13, 17 mm
Catalog # ECHLRMAG0043
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Magnification:4×
- Description:Plan achromat phase, N.A. 0,13
- Seq:29
- Working distance:17 mm