Orders LinkContactUsLinkComponent
Labels, hazard warning
Labels, hazard warning
Hazard labels: drop, PIC W008-TRI 050-PE-ROLL/1, Polyester, Black on yellow, 50×36 mm, 250 labels/roll
Catalog # BRDY827500
Supplier:   Brady
Labels, hazard warning
Catalog # BRDY827500
Supplier:  Brady
Labels, hazard warning
Labels, hazard warning
Hazard labels: drop, PIC W008-TRI 050-PE-ROLL/1, Polyester, Black on yellow, 50×36 mm, 250 labels/roll
Price
142,10 €
Log in for availability
1 * 1 ROU
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
  • Colour:
    Black on yellow
  • Description:
    Hazard labels: drop
  • Dimensions:
    50×36 mm
  • Material:
    Polyester
  • Model:
    PIC W008-TRI 050-PE-ROLL/1
  • Packed:
    250 labels/roll