Labels, hazard warning
Hazard labels: explosive material, PIC W002-TRI 025-PE-CRD/1, Polyester, Black on yellow, 25×18 mm, 250 labels/roll
Catalog # BRDY816661
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: explosive material
- Dimensions:25×18 mm
- Material:Polyester
- Model:PIC W002-TRI 025-PE-CRD/1
- Packed:250 labels/roll