Autoclavable goggles
Cleanroom goggles, Anti-UV, autoclavable, indirectly vented, Clear PC, Blue
N° de catalogue 111-2429
Fournisseur: UNIVET
Jump to:
Product Details & Documents
The autoclavable goggles are designed for use in rooms with controlled contaminations or in aseptic conditions.
- Available with direct or indirect ventilation
- Elastic band in anti-static silicone with quick and smooth system
- RX insert
- Goggles with polycarbonate lens reusable up to 40 autoclave
The soft structure supports multiple cycles of steam, gamma or beta sterilisation without perceptible changes. The autoclavable goggles has a polycarbonate lens with anti-fog coating AF121 and anti-scratch.
Certifications: EN 166.
Delivery information: Available with correction insert and replacement lens.
Documents
Specifications
- Type:Anti-UV, autoclavable, indirectly vented
- Colour:Blue
- Description:Cleanroom goggles
- Seq:04
- Lenses:Clear PC
Accessories
Product Information
- N° de catalogueDésignationDisponibilitéPrix
Product Information
- N° de catalogueDésignationDisponibilitéPrix
Product Information
- N° de catalogueDésignationDisponibilitéPrix
Product Information
- N° de catalogueDésignationDisponibilitéPrix