Order Entry
Belgium
Orders LinkContactUsLinkComponent
Washers, BOLA

Washers, BOLA

for tubing 0,8 mm I-Ø/1,6 mm O-Ø, PA
Catalog # BOHLF728-08
Supplier:   Bohlender
Washers, BOLA
Catalog # BOHLF728-08
Supplier:  Bohlender
Washers, BOLA
Washers, BOLA

for tubing 0,8 mm I-Ø/1,6 mm O-Ø, PA

Price
6,10 €
Log in for availability
1 * 10 ST
Product Details & Documents

PA

  • Temperature resistance: from –40 to +100 °C

For stabilising the flange and avoiding damages when tightening the tube end fitting.

Specifications
  • Description:
    for tubing 0,8 mm I-Ø/1,6 mm O-Ø
  • Material:
    PA
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.