Vented QTray with cover, polystyrene, 240 x 240 x 20 mm 20 per case 1 * 20 items
Catalog # MLDVX6023
Supplier: Molecular Devices
Vented QTray with cover, polystyrene, 240 x 240 x 20 mm 20 per case 1 * 20 items
Catalog # MLDVX6023
Supplier: Molecular Devices
Frequently Bought Together
VWR®, Universal Pipette Tips
Supplier: VWR Collection
Disposable scalpels, sterile
Supplier: SWANN MORTON
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Beakers, Low Form
Supplier: VWR Collection
MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
Potassium dihydrogen phosphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Inoculating needles and loops, Nunc™
Supplier: Thermo Fisher Scientific
Dextrin
Supplier: Thermo Fisher Scientific
Labels, hazard warning
Supplier: Brady
QIAQUICK NUCLEOTIDE REMOVAL KIT (250) 1 * 250 items
Supplier: QIAGEN
Universal container with screw cap, PS, 30 ml, STERILIN®
Supplier: STERILIN