Orders LinkContactUsLinkComponent
Multi management software, APT-COM™ 4
Multi management software, APT-COM™ 4
APT-COM™ 4 GLP-Edition, for working under GLP-compliant conditions. Measured values are documented in a tamper-proof way in line with the requirements of FDA Regulation 21 CFR 11, UF V
Catalog # BIND9053-0042
Supplier:   Binder
Multi management software, APT-COM™ 4
Catalog # BIND9053-0042
Supplier:  Binder
Multi management software, APT-COM™ 4
Multi management software, APT-COM™ 4
APT-COM™ 4 GLP-Edition, for working under GLP-compliant conditions. Measured values are documented in a tamper-proof way in line with the requirements of FDA Regulation 21 CFR 11, UF V
Price
5 060,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This software provides management, logging, programming, documentation options, and much more. With features ranging from GLP-compliant logging to convenient program creation and management of up to 100 units, the APT-COM™ 4 can be used in all industries and makes complex work processes much easier.
  • Central overview of all units in both graphic and table form
  • Record management (creating, deleting, archiving)
  • Choice of five menu languages: DE, EN, FR, IT, ES
  • Graphical/numerical program editor
  • Reminders can be set via timer function
  • Documentation of measured values
  • Export of measured values in CSV and PDF format
BASIC Edition: For simple recording and documentation requirements with up to five networked units.
PROFESSIONAL Edition: Convenient unit and user management which builds on the BASIC edition. Suitable for networking up to 100 units.
GLP Edition: For working under GLP-compliant conditions. Measured values are documented in a tamperproof way in line with the requirements of FDA Regulation 21 CFR 11.
Specifications
  • Description:
    APT-COM™ 4 GLP-Edition, for working under GLP-compliant conditions. Measured values are documented in a tamper-proof way in line with the requirements of FDA Regulation 21 CFR 11
  • Seq:
    03
  • For:
    UF V