ContactUsLinkComponent
1.0ml External Thread Co-Molded Jacket Tube with 2D code on Tube Base & Linear Barcode, HRN on side, with Screw Cap, bulk 960 tubes per case 1 * 1 items
N° de catalogue BCIS68-1003-10
Fournisseur:  BIOCISION
1.0ml External Thread Co-Molded Jacket Tube with 2D code on Tube Base & Linear Barcode, HRN on side, with Screw Cap, bulk 960 tubes per case 1 * 1 items
N° de catalogue BCIS68-1003-10
Fournisseur:  BIOCISION
Frequently Bought Together