Order Entry
Belgium
Orders LinkContactUsLinkComponent
Assistent® Pipette brushes

Assistent® Pipette brushes

90 mm, 30 mm, 300 mm
Catalog # HECH91120000010
Assistent® Pipette brushes
Catalog # HECH91120000010
Assistent® Pipette brushes
Assistent® Pipette brushes

90 mm, 30 mm, 300 mm

Price
238,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Straight brushes in different sizes.

Documents

Specifications
  • Brush head Ø:
    30 mm
  • Seq:
    05
  • Total length:
    300 mm
  • Brush head length:
    90 mm