Order Entry
Belgium
Orders LinkContactUsLinkComponent
Assistent® Pipette brushes

Assistent® Pipette brushes

70 mm, 15 mm, 250 mm
Catalog # HECH91120000007
Assistent® Pipette brushes
Catalog # HECH91120000007
Assistent® Pipette brushes
Assistent® Pipette brushes

70 mm, 15 mm, 250 mm

Price
225,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Straight brushes in different sizes.

Documents

Specifications
  • Brush head Ø:
    15 mm
  • Seq:
    03
  • Total length:
    250 mm
  • Brush head length:
    70 mm