Orders LinkContactUsLinkComponent
Labels for BradyJet J2000 Lab printer
Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-59-7425J, PP, White, J2000-Lab, 25×12 mm, 2000 labels/roll
Catalog # BRDY150006
Supplier:   Brady
Labels for BradyJet J2000 Lab printer
Catalog # BRDY150006
Supplier:  Brady
Labels for BradyJet J2000 Lab printer
Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-59-7425J, PP, White, J2000-Lab, 25×12 mm, 2000 labels/roll
Price
164,60 €
Log in for availability
1 * 1 ROU
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
  • Designed to stay attached and remain legible
  • Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Specifications
  • Printer model:
    J2000-Lab
  • Type:
    B-7425J
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    25×12 mm
  • Material:
    PP
  • Seq:
    04
  • Model:
    J20-59-7425J
  • Packed:
    2000 labels/roll
  • Cat. No.:
    BRDY150006