ContactUsLinkComponent
Disposable mop Clino One Way Ultra PW, prewashed, for use in cleanrooms ISO 5-8 / GMP grade C-D, non-sterile, double-packed 1 * 5 items
Catalog # 141-1456
Disposable mop Clino One Way Ultra PW, prewashed, for use in cleanrooms ISO 5-8 / GMP grade C-D, non-sterile, double-packed 1 * 5 items
Catalog # 141-1456