ContactUsLinkComponent
Flexible discharge tube for genius / simplex 2, 5 and 10 ml Coiled, made of FEP, approx. 80 cm length, with handle and recirculation valve made of PTFE. Includes handle and assembly instructions. Not suitable for hydrofluoric acid (HF) 1 * 1 items
Catalog # VITL1678132
Supplier:  VITLAB
Flexible discharge tube for genius / simplex 2, 5 and 10 ml Coiled, made of FEP, approx. 80 cm length, with handle and recirculation valve made of PTFE. Includes handle and assembly instructions. Not suitable for hydrofluoric acid (HF) 1 * 1 items
Catalog # VITL1678132
Supplier:  VITLAB
Frequently Bought Together