VWR® Evaporating Flasks
1000 ml, 29/32 NS
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance and high resistance to thermal shock
- White labelling area and imprinted catalogue number
These pear-shaped rotary evaporating flasks are produced from thick walled borosilicate glass tubing. With standard ground socket, suitable for all standard rotary evaporators.
Specifications
- Socket size:29/32 NS
- Capacity:1000 ml
- Seq:04