Orders LinkContactUsLinkComponent
VWR® Evaporating Flasks
VWR® Evaporating Flasks
250 ml, 29/32 NS
Catalog # 531-1100
Supplier:   VWR Collection
VWR® Evaporating Flasks
Catalog # 531-1100
Supplier:  VWR Collection
VWR® Evaporating Flasks
VWR® Evaporating Flasks
250 ml, 29/32 NS
Price
34,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • White labelling area and imprinted catalogue number
These pear-shaped rotary evaporating flasks are produced from thick walled borosilicate glass tubing. With standard ground socket, suitable for all standard rotary evaporators.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    250 ml
  • Seq:
    02