ContactUsLinkComponent
Electroporation cuvette plus 4mm gap 800µl yellow sterile 1 * 10 items
Catalog # BTXI45-0136
Supplier:  BTX
Electroporation cuvette plus 4mm gap 800µl yellow sterile 1 * 10 items
Catalog # BTXI45-0136
Supplier:  BTX
Frequently Bought Together