ContactUsLinkComponent
A Akr7a2 (Aflatoxin B1 aldehyde reductase member 2) ELISA with a senstivity of 46.875pg/ml and a range of 78.125-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8279
Supplier:  G-Biosciences
A Akr7a2 (Aflatoxin B1 aldehyde reductase member 2) ELISA with a senstivity of 46.875pg/ml and a range of 78.125-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8279
Supplier:  G-Biosciences
Frequently Bought Together