ContactUsLinkComponent
Quantifies changes in mitochondrial membrane potential of live cells via microplate fluorometry, fluorescence microscopy and flow cy tometry. TMRE is a cell permeable, positively charged red-orange dye that accumulates in active mitochondria due to t 1 * 500 Tests
Catalog # GENO786-1314
Supplier:  G-Biosciences
Quantifies changes in mitochondrial membrane potential of live cells via microplate fluorometry, fluorescence microscopy and flow cy tometry. TMRE is a cell permeable, positively charged red-orange dye that accumulates in active mitochondria due to t 1 * 500 Tests
Catalog # GENO786-1314
Supplier:  G-Biosciences
Frequently Bought Together