Orders LinkContactUsLinkComponent
Sponge sticks
Sponge sticks
Media type: Letheen broth
Catalog # MMMPSSL10LET
Supplier:   Neogen
Sponge sticks
Catalog # MMMPSSL10LET
Supplier:  Neogen
Sponge sticks
Sponge sticks
Media type: Letheen broth
Price
245,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
3M™ Sponge-Stick in sample bag with 10 ml D/E neutralising broth uses simplified packaging to minimise handling and preparation, and reduces waste.
  • Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge
  • Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing
  • Simplified packaging minimises handling and preparation and reduces waste
Better testing begins with better samples and better samples are obtained when you use the best collection tools. 3M™ Sponge-Stick allows sampling without directly handling the sponge. It makes it easier to reach into drains, pipes and around equipment. Biocide-free cellulose sponge maintains organism viability. These are convenient and ready-to-use with consistent, reliable results every time.
Delivery information: Supplied in wet or dry formats to fit many applications.
Documents
Specifications
  • Description:
    Media type: Letheen broth
  • Seq:
    04
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.