Robust Temperature Data Logger, EBI 12-T100
Product Details & Documents
Discover the EBI 12-T100 from ebro: a robust temperature data logger, ideal for independent routine checks in medicine, pharmaceuticals and production. Sterilisable and with a large measuring range for the optimal quality assurance.
- Wide measuring range, –90 to +150 °C
- High-quality PT1000 sensor (class A)
- Robust IP 68 protection class: waterproof and vapour-tight
- Supplied with calibration certificate
- Sterilisable data logger
The EBI 12-T100 from ebro is a high-performance temperature data logger. Its impressive measuring range of –90 to +150 °C and the high-quality PT1000 sensor (class A) offer maximum precision and enable versatile applications from dry ice to steam sterilisers.
Thanks to its robust IP 68 protection class, it is waterproof and vapour-tight. Each device is supplied with a calibration certificate to ensure maximum accuracy.
Dimensions: 48Ø×24H mm.
Factory calibration certificate: –20, 0, 60 and 134 °C.
Certifications: (EG) 1935/2004.
Documents
Manual Robust Temperature Data Logger, EBI 12-T100, Ebro Manual Temperature and Pressure Data Logger EBI 12-EX.pdf
Manual
Professional Data Logger for Validation, Routine Control and Process Monitoring.pdf Proffessional Dataloggers Validation and Process Monitoring EBI11-12.pdf Validation and Process Monitoring Food.pdf Validation and Process Monitoring.pdf
Additional Product Details
Professional Dataloggers
Product Literature
Robust Temperature Data Logger, EBI 12-T100
Application Notes
Specifications
- Range:EX-areas: –40…+85 °C; T: –90…+150 °C
- Resolution:0,01 °C
- Description:EBI 12-T100-EX, temperature data logger for hazardous areas
- Seq:3
- Accuracy:±0,5 °C (–90…–40 °C); ±0,2 °C (–40…0 °C); ±0,1 °C (0…120 °C); ±0,05 °C (120…140 °C); ±0,1 °C (140…150 °C)
- Cat. No.:EBRO1340-6600-EX
- Recording intervals:250 ms - 24 hrs.
- Protection class:IP 68
- Data memory - No. of readings:100320
- Model:EBI 12-T100-EX
- Batteries:Lithium battery, 3,6 V
- Weight:90 g
- Sensor:PT1000, class A
Complementary Products
Product Information
- Catalog #AccuracyDescriptionRangeResolutionAvailabilityPrice
- EBRO1340-6601-0100±0,3 °C (0…105 °C)
±0,5 °C (–90 …–40 °C)EBI 12-T101, data logger for washer and disinfecter; calibration points –20 °C, 0 °C and +60 °CT: –90…+105 °C0,01 °CLog in for availability1 * 1 ST - 1 650,00 € - EBRO1340-6658±0,1 (0…140 °C)
±20 mbarEBI 12-TP237, temperature and pressure data logger, calibration points 0 °C, +60 °C and +134 °C; 100 mbar and 3100 mbar (bei +25 °C); 3100 mbar (at +134 °C)T: 0…140 °C
P: 1…4000 mbar0,01 °C
1 mbarLog in for availability1 * 1 ST - 2 440,00 € - EBRO1340-6601±0,5 °C (–90…–40 °C); ±0,3 °C (–40…105 °C)EBI 12-T101, Temperature data logger, pharmaceutical, medical and food applications (factory calibration certificate: –20 °C, 0 °C and +60 °C)T: –90…+105 °C0,01 °CLog in for availability1 * 1 ST - 1 790,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Battery replacement set EBI 12; 3× battery, 6× O-ringBattery replacement set EBI 12; 3× battery, 6× O-ring620-1693Log in for availability1 * 1 ST - 194,00 €
Product Information
- Catalog #AvailabilityPrice
- Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12EBRO1100-0130Log in for availability1 * 1 ST - 277,00 €
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



