[EN]BLITZ 10X KINETIC BUFFER PROCLIN 1X
Catalog # PALF18-1105
Supplier: PALL FORTEBIO
Frequently Bought Together
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
BIOSENSOR / ANTI-MOUSE FC CAPTURE (AMC) PACK 1 * 5 items
Supplier: PALL FORTEBIO
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
[EN]BIOSENSOR - HIGH PRECISION STREP (SA
Supplier: PALL FORTEBIO
NI-NTA(NTA) BIOSENSORS FIVE TRAYS OF 96 1 * 1 KIT
Supplier: PALL FORTEBIO