ContactUsLinkComponent
MiniLab restriction digest basics. Includes GreenGel cups (10), TBE concentrate (100ml), 0,7ml multi-colour microcentrifuge tubes (5 5 tubes), universal fit micropipette tips, MiniOne universal DNA Marker (130µl), five pre-digested DNA samples (PCR f 1 * 1 KIT
Catalog # EMBIM6050
Supplier:  EMBITEC
MiniLab restriction digest basics. Includes GreenGel cups (10), TBE concentrate (100ml), 0,7ml multi-colour microcentrifuge tubes (5 5 tubes), universal fit micropipette tips, MiniOne universal DNA Marker (130µl), five pre-digested DNA samples (PCR f 1 * 1 KIT
Catalog # EMBIM6050
Supplier:  EMBITEC
Frequently Bought Together