ContactUsLinkComponent
MiniOne PCR system. Includes thermal cycler with 16 sample well capacity. Fast heating & cooling based on peltier-effect thermoelect ric, heated lid, wireless connectivity to iPad or Android mobile tablets. System comes with a 12V 100W power supply ( 1 * 1 items
Catalog # EMBIM4000
Supplier:  EMBITEC
MiniOne PCR system. Includes thermal cycler with 16 sample well capacity. Fast heating & cooling based on peltier-effect thermoelect ric, heated lid, wireless connectivity to iPad or Android mobile tablets. System comes with a 12V 100W power supply ( 1 * 1 items
Catalog # EMBIM4000
Supplier:  EMBITEC
Frequently Bought Together