5-100mm Path Cylindrical Cell Holder for GENESYS 100 series. Holds one cylindrical longpath cell 5 - 100mm pathlength. Comes assem bled in a removable sample compartment liner. 1 * 1 items
Catalog # 634-1096
Supplier: Thermo Fisher Scientific
Frequently Bought Together