Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sampling spoons, SteriWare®

Sampling spoons, SteriWare®

Sampling spoons, non sterile, 300 mm, 10 ml
N° de catalogue SAMP8293H-10
Fournisseur:   Sampling Systems
Sampling spoons, SteriWare®
N° de catalogue SAMP8293H-10
Fournisseur:  Sampling Systems
Sampling spoons, SteriWare®
Sampling spoons, SteriWare®

Sampling spoons, non sterile, 300 mm, 10 ml

Prix
290,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

SteriWare® Single use long handled spoons are ideal for reaching into large containers or difficult to reach places. It can be used straight from the bag and manufactured under cleanroom conditions so they can be used without costly cleaning.

  • Made from FDA Conforming HDPE
  • Pre-sterilised versions are available
  • Moulded and suitable for powders and liquids
  • Fully validated sterilisation procedure (SAL 10⁻⁶)
  • Full batch traceability

Certifications: FDA, EU 10/2011 and EC 1935/2004 conformity; BSE/TSE free.

Packaging: Individually bagged in a heat-sealed laminate bag.

Specifications
  • Capacity:
    10 ml
  • Description:
    Sampling spoons, non sterile
  • Length:
    300 mm
  • Seq:
    06
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.