Orders LinkContactUsLinkComponent
Laboratory software, Labworldsoft 6
Laboratory software, Labworldsoft 6
Labworldsoft® 6 Pro, with access to the entire device library, All configurations with any number of devices can be controlled at the same time
Catalog # IKAA20017366
Supplier:   IKA
Laboratory software, Labworldsoft 6
Catalog # IKAA20017366
Supplier:  IKA
Laboratory software, Labworldsoft 6
Laboratory software, Labworldsoft 6
Labworldsoft® 6 Pro, with access to the entire device library, All configurations with any number of devices can be controlled at the same time
Price
14 820,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Labworldsoft 6 is a laboratory software to control lab devices, automate processes, and to document your measurements and results. The software provides access to a huge library of different laboratory devices.
  • Easy recording of various physical parameters
  • Function modules to create time triggered or event triggered configurations
  • Communication through RS232, USB, Bluetooth or network
  • Supports formats .csv, .xslx and ASCII
  • Compatibility with Windows 7, 8 and 10
Labworldsoft does not integrate only IKA laboratory devices, but supports several other device from different manufacturers as well. You stay flexible in the choice of your lab devices you want to use and profit from the whole device range of the laboratory world.
Delivery information: Includes one year of support with new versions of Labworldsoft 6.
Specifications
  • Description:
    Labworldsoft® 6 Pro, with access to the entire device library
  • Seq:
    02
  • For:
    All configurations with any number of devices can be controlled at the same time