SCREWCAP WHITE PP W/CONE-SEAL 28MM 1 * 3.600 items
Catalog # RIXI8-2000-28-W
Fournisseur: RIXIUS
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Fournisseur: HEATHROW
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
VWR® Facial Tissues
Fournisseur: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
VWR®, Nitrile Gloves
Fournisseur: VWR Collection
pH indicator strips, MQuant®, Supelco®
Fournisseur: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Fournisseur: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Fournisseur: VWR Collection
VWR®, Transfer Pipettes, High Performance
Fournisseur: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Fournisseur: VWR Collection