ContactUsLinkComponent
Photometer with 280nm light source: Fiber optic cables, 2 optical cables couplers to connect to flow cell, 24VDC power supply 1 * 1 items
Catalog # SPECP-ACCD-D-U280
Photometer with 280nm light source: Fiber optic cables, 2 optical cables couplers to connect to flow cell, 24VDC power supply 1 * 1 items
Catalog # SPECP-ACCD-D-U280
Frequently Bought Together