TROCKENMITTEL CALCIUMCHLORID, =98 % GETROCKNET, PULVER, REGENERIERUNG BEI 250°C 1 * 500 g
ROTHCN93.1
TROCKENMITTEL CALCIUMCHLORID, =98 % GETROCKNET, PULVER, REGENERIERUNG BEI 250°C 1 * 500 g
ROTHCN93.1
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection