VISCOSITY STANDARD S3 AP (L) 1 * 100 mL
Catalog # PAAR100034
Supplier: Anton Paar
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
di-Ammonium oxalate solution Reag. Ph. Eur. 1005901
Supplier: VWR Chemicals
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
VWR® ARISTAR®, Yttrium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
C TECHNOLOGIES VPE MEASUREMENT STANDAR
Supplier: MARKET SOURCE PART PROCESS
ISE standards
Supplier: Hach
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Triethyl Citrate, Reference Standard
Supplier: USP
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Conductivity standards, Potassium chloride, IUPAC
Supplier: Hach
Turbidity standards, Stablcal®
Supplier: Hach