Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 25 - 27%, NORMATOM® for trace analysis

Ammonia solution 25 - 27%, NORMATOM® for trace analysis

1 L
Catalog # 85693.290
Corrosive
Irritant
Supplier:   VWR Chemicals
Ammonia solution 25 - 27%, NORMATOM® for trace analysis
Catalog # 85693.290
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution 25 - 27%, NORMATOM® for trace analysis
Ammonia solution 25 - 27%, NORMATOM® for trace analysis

1 L

CAS Number:  1336-21-6
Price
172,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay (NH3):
    25 - 27% w/w
  • Colouration:
    max. 10 APHA
  • Cl (Chloride):
    max. 0.5 ppm
  • PO4 (Phosphate):
    max. 0.01 ppm
  • SO4 (Sulphate):
    max. 1 ppm
  • Ag (Silver):
    max. 0.5 ppb
  • Al (Aluminum):
    max. 1 ppb
  • As (Arsenic):
    max. 1 ppb
  • Au (Gold):
    max. 0.5 ppb
  • Ba (Barium):
    max. 0.1 ppb
  • Be (Beryllium):
    max. 0.1 ppb
  • Bi (Bismuth):
    max. 0.1 ppb
  • Ca (Calcium):
    max. 1 ppb
  • Cd (Cadmium):
    max. 0.5 ppb
  • Ce (Cerium):
    max. 0.1 ppb
  • Co (Cobalt):
    max. 0.5 ppb
  • Cr (Chromium):
    max. 0.5 ppb
  • Cs (Cesium):
    max. 0.1 ppb
  • Cu (Copper):
    max. 0.5 ppb
  • Dy (Dysprosium):
    max. 0.1 ppb
  • Er (Erbium):
    max. 0.1 ppb
  • Eu (Europium):
    max. 0.1 ppb
  • Fe (Iron):
    max. 1 ppb
  • Ga (Gallium):
    max. 0.1 ppb
  • Gd (Gadolinium):
    max. 0.1 ppb
  • Ge (Germanium):
    max. 0.1 ppb
  • Hf (Hafnium):
    Information Only
  • Hg (Mercury):
    max. 0.2 ppb
  • Ho (Holmium):
    max. 0.1 ppb
  • In (Indium):
    max. 0.1 ppb
  • K (Potassium):
    max. 1 ppb
  • La (Lanthanum):
    max. 0.1 ppb
  • Li (Lithium):
    max. 0.1 ppb
  • Lu (Lutetium):
    max. 0.1 ppb
  • Mg (Magnesium):
    max. 1 ppb
  • Mn (Manganese):
    max. 0.5 ppb
  • Mo (Molybdenum):
    max. 0.5 ppb
  • Na (Sodium):
    max. 1 ppb
  • Nb (Niobium):
    max. 0.1 ppb
  • Nd (Neodymium):
    max. 0.1 ppb
  • Ni (Nickel):
    max. 0.5 ppb
  • Pb (Lead):
    max. 0.1 ppb
  • Pd (Palladium):
    Information Only
  • Pr (Praseodymium):
    max. 0.1 ppb
  • Pt (Platinum):
    Information Only
  • Rb (Rubidium):
    max. 0.1 ppb
  • Re (Rhenium):
    Information Only
  • Rh (Rhodium):
    max. 0.5 ppb
  • Ru (Ruthenium):
    Information Only
  • Sb (Antimony):
    max. 0.5 ppb
  • Sc (Scandium):
    max. 0.1 ppb
  • Se (Selenium):
    max. 1 ppb
  • Sm (Samarium):
    max. 0.1 ppb
  • Sn (Tin):
    max. 0.5 ppb
  • Sr (Strontium):
    max. 0.1 ppb
  • Tb (Terbium):
    max. 0.1 ppb
  • Te (Tellurium):
    max. 0.1 ppb
  • Th (Thorium):
    max. 0.1 ppb
  • Ti (Titanium):
    max. 0.5 ppb
  • Tl (Thallium):
    max. 0.1 ppb
  • Tm (Thulium):
    max. 0.1 ppb
  • U (Uranium):
    max. 0.1 ppb
  • V (Vanadium):
    max. 0.5 ppb
  • W (Tungsten):
    max. 0.1 ppb
  • Y (Yttrium):
    max. 0.1 ppb
  • Yb (Ytterbium):
    max. 0.1 ppb
  • Zn (Zinc):
    max. 0.5 ppb
  • Zr (Zirconium):
    max. 0.1 ppb

Specifications

  • Pk:
    1 L

Specifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.