SINGLE USE PRESSURE SENSOR, NON-STERILE, POLYSULPHONE, LUER INLET/OULET 1 * 1 items
Catalog # PENDPREPS-N-000
Supplier: PENDOTECH
SINGLE USE PRESSURE SENSOR, NON-STERILE, POLYSULPHONE, LUER INLET/OULET 1 * 1 items
Catalog # PENDPREPS-N-000
Supplier: PENDOTECH
Frequently Bought Together
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Supplier: Avantor Fluid Handling
Hose Clamps, Adjustable, Stainless Steel
Supplier: Avantor Fluid Handling
Disposable scalpels, sterile
Supplier: SWANN MORTON
Masterflex® Union Fittings, Hose Barb, Straight, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Luer to Luer, Polycarbonate, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® One-Way Luer Check Valve Fittings, Luer, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
HelixMark® Peristaltic Pump Tubing
Supplier: Freudenberg Medical Europe GmbH
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Lab markers
Supplier: MICRONOVA
Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hosebarb to NPT(M), Straight, Polyproylene, Avantor®
Supplier: Avantor Fluid Handling