ContactUsLinkComponent
RS 232 with 25-pin connector communication kit, includes communications board and wiring harnesses. Communicates information from Purifier Logic+ Biosafety Cabinets to user-supplied and operated computer. EA 1 * 1 KIT
Catalog # LBCP3858311
Supplier:  LABCONCO
RS 232 with 25-pin connector communication kit, includes communications board and wiring harnesses. Communicates information from Purifier Logic+ Biosafety Cabinets to user-supplied and operated computer. EA 1 * 1 KIT
Catalog # LBCP3858311
Supplier:  LABCONCO
Frequently Bought Together