ContactUsLinkComponent
Accredited temperature and humidity Calibration Certificate for AiroSensor. Values: -10C, 20C, 50C and 30%, 80% RH. Batch size of 10 AiroSensors or more. 1 * 1 items
Catalog # SENA0630125977224
Supplier:  SenseAnywhere
undefined
Accredited temperature and humidity Calibration Certificate for AiroSensor. Values: -10C, 20C, 50C and 30%, 80% RH. Batch size of 10 AiroSensors or more. 1 * 1 items
Catalog # SENA0630125977224
Supplier:  SenseAnywhere
Frequently Bought Together