ContactUsLinkComponent
Slides 8 wells 6mm diameter red numbered 1 * 50 items
Catalog # MARI1216754
Supplier:  MARIENFELD
Slides 8 wells 6mm diameter red numbered 1 * 50 items
Catalog # MARI1216754
Supplier:  MARIENFELD
Frequently Bought Together