Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mouse Recombinant FGF-Basic (from <i>E. coli</i>)

Mouse Recombinant FGF-Basic (from E. coli)

Vial, 1 mg
Catalog # SHBT200-12AF-1MG
Mouse Recombinant FGF-Basic (from <i>E. coli</i>)
Catalog # SHBT200-12AF-1MG
CAS Number:  
Mouse Recombinant FGF-Basic (from E. coli)

Vial, 1 mg

CAS Number:  
Price
1 680,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.

  • High quality
  • Low endotoxin
  • Affordable

 : This product is for research use only and is not for use in humans.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    16,5 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Protein function:
    Cytokine
  • Gene ID:
    P15655
  • Formulation:
    10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Mouse
  • Protein/peptide name:
    FGF-Basic (AF)
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Biological activity:
    Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT200-12AF-1MG
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    Y

Specifications