Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Human Recombinant FGF-Basic 154 (from E. coli)

Vial, 10 µG
Catalog # SHBT100-146AF-10UG
Human Recombinant FGF-Basic 154 (from <i>E. coli</i>)
Catalog # SHBT100-146AF-10UG
CAS Number:  
Human Recombinant FGF-Basic 154 (from <i>E. coli</i>)
Human Recombinant FGF-Basic 154 (from E. coli)

Vial, 10 µG

CAS Number:  
Price
82,30 €
Log in for availability
1 * 1 EA
Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.

  • High quality
  • Low endotoxin
  • Affordable

 : This product is for research use only and is not for use in humans.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    10 µG
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    17,3 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Protein function:
    Cytokine
  • Gene ID:
    P09038
  • Formulation:
    10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Human
  • Protein/peptide name:
    FGF-Basic 154 (AF)
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
  • Biological activity:
    Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT100-146AF-10UG
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    Y

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.