Order Entry
Belgium
Orders LinkContactUsLinkComponent
Accessories for REVOLVE

Accessories for REVOLVE

Stage insert, glass slides
Catalog # ECHLHLDR0002
Supplier:   DISCOVER ECHO
Accessories for REVOLVE
Catalog # ECHLHLDR0002
Supplier:  DISCOVER ECHO
Accessories for REVOLVE

Stage insert, glass slides

Price
147,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Stage insert, glass slides
  • Seq:
    04
  • Cat. No.:
    ECHLHLDR0002
  • Observation method:
    Brightfield and phase contrast
  • Output:
    USB
  • Electrical:
    163 - 240 VA (50/60 Hz)
  • Camera:
    75 MP colour
  • Display:
    12,9” iPad Pro® Retina® display, 2732×2048 (264 pixels/inch)
  • Sensor:
    CMOS