Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat ELWD phase objective, 2,2 mm
Catalog # ECHLJMAG0409
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0409
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat ELWD phase objective, 2,2 mm
Price
1 530,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    40×
  • Description:
    Achromat ELWD phase objective
  • Seq:
    20
  • Working distance:
    2,2 mm