Objectives for REVOLVE 3M microscope
Achromat phase objective, 0,6 mm
Catalog # ECHLJMAG0408
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Specifications
- Magnification:40×
- Description:Achromat phase objective
- Seq:19
- Working distance:0,6 mm