Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Fluorite objective, 0,51 mm
Catalog # 630-3061
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3061
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite objective, 0,51 mm
Price
1 490,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    40×
  • Description:
    Fluorite objective
  • Seq:
    10
  • Working distance:
    0,51 mm