Objectives for REVOLVE 3M microscope
Achromat objective, 1,2 mm
Catalog # ECHLJMAG0206
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifications
- Magnification:20×
- Description:Achromat objective
- Seq:12
- Working distance:1,2 mm