Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Fluorite ELWD objective, 6,6 - 7,8 mm
Catalog # 630-3055
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3055
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite ELWD objective, 6,6 - 7,8 mm
Price
2 110,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    20×
  • Description:
    Fluorite ELWD objective
  • Seq:
    23
  • Working distance:
    6,6 - 7,8 mm