Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat ELWD phase objective, 9,8 mm
Catalog # ECHLJMAG0106
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0106
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat ELWD phase objective, 9,8 mm
Price
632,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    10×
  • Description:
    Achromat ELWD phase objective
  • Seq:
    09
  • Working distance:
    9,8 mm