Orders LinkContactUsLinkComponent
Colour coded inserts, CapInsert™
Colour coded inserts, CapInsert™
Blue
Catalog # SIMPM957B
Supplier:   Simport Scientific
Colour coded inserts, CapInsert™
Catalog # SIMPM957B
Supplier:  Simport Scientific
Colour coded inserts, CapInsert™
Colour coded inserts, CapInsert™
Blue
Price
21,80 €
Log in for availability
1 * 200 ST
Product Details & Documents
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
  • Manually press fit and locks into place on top of closure
Specifications
  • Colour:
    Blue
  • Seq:
    01