ContactUsLinkComponent
Immersion coolers (Dephlegmators) Preciso, cone ST 19/26, effective length: 100 mm, total length: 185 mm, with 2x GL14 1 * 1 items
Catalog # WITG1.786.219.S
Supplier:  witeg Labortechnik
Immersion coolers (Dephlegmators) Preciso, cone ST 19/26, effective length: 100 mm, total length: 185 mm, with 2x GL14 1 * 1 items
Catalog # WITG1.786.219.S
Supplier:  witeg Labortechnik
Frequently Bought Together